Difco™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
212489
212488
76628-756EA
2538.74
USD
76628-756
76628-744
Difco™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology
Additives and Supplements for Microbiology
Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, this supplement offers an alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO.
- High concentrations of vitamins and carbohydrates
- An enzymatic digest of soy