2-Fluoro-4-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4373F-100G PC4373F-25G PC4373F-5G
76626-464EA 219.02 USD
76626-464 76626-466 76626-468
2-Fluoro-4-(trifluoromethyl)benzonitrile 97%
2-Fluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
Boiling Pt: 178…180 °C
Density: 1.358 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00061279
CAS Number: 146070-34-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR