2-Aminonicotinonitrile

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine

OR8628-1G
76625-486EA 20.87 USD
76625-486
2-Aminonicotinonitrile
2-Aminonicotinonitrile
Formula: C₆H₅N₃
MW: 119.13 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00128867
CAS Number: 24517-64-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR