α,4-Dichlorotoluene 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

OR3764-250G
76625-152EA 93.73 USD
76625-152 76625-154 76625-156
α,4-Dichlorotoluene 99%
α,4-Dichlorotoluene
Formula: C₇H₆Cl₂
Boiling Pt: 216…222 °C
Melting Pt: 26…30 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR