2-Ethynyltoluene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Ethynyl-2-methylbenzene, 2-Methylphenylacetylene

OR3480-1G OR3480-25G OR3480-5G
76625-140EA 84.54 USD
76625-140 76625-142 76625-144
2-Ethynyltoluene
2-Ethynyltoluene
Formula: C₉H₈
MW: 116.1598 g/mol
Boiling Pt: 50…60 °C/30mmHg
Density: 0.922 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427202
CAS Number: 766-47-2
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR