3-Hydroxythiophenol

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Hydroxybenzene-1-thiol, 3-Hydroxybenzenethiol, 3-Hydroxythiophenol, 1-Hydroxy-3-mercaptobenzene, 3-Mercaptophenol, m-Hydroxythiophenol, 3-Hydroxyphenyl mercaptan

OR0777-100G OR0777-25G OR0777-5G
76624-662EA 441.08 USD
76624-662 76624-664 76624-666
3-Hydroxythiophenol
3-Hydroxythiophenol
Formula: C₆H₆OS
MW: 126.18
Boiling Pt: 242…243 °C
Melting Pt: 16.5
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317829
CAS Number: 40248-84-8
UN: 3334
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR