ω-Bromo-3-fluoroacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4059W-100G PC4059W-25G PC4059W-5G
76623-520EA 542.94 USD
76623-520 76623-522 76623-524
ω-Bromo-3-fluoroacetophenone 97%
ω-Bromo-3-fluoroacetophenone
Formula: C₈H₆BrFO
Boiling Pt: 76 °C/0.09mmHg
Melting Pt: 32…37 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00109756
CAS Number: 53631-18-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR