3-(4-Methylpiperazin-1-yl)aniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR9659-1G OR9659-5G
76622-024EA 20.87 USD
76622-024 76622-026
3-(4-Methylpiperazin-1-yl)aniline 98%
3-(4-Methylpiperazin-1-yl)aniline
Formula: C₁₁H₁₇N₃
Melting Pt: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD08435857
CAS Number: 148546-99-0
UN: 3263
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR