α-Bromo-4-iodotoulene 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR8750-100G
OR8750-25G
OR8750-5G
76621-926EA
231.24
USD
76621-926
76621-928
76621-930
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene
Formula:
C₇H₆BrI Boiling Pt: 140 °C/15mmHg Melting Pt: 78…82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00209656 CAS Number: 16004-15-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...