3-Chlorobenzenesulfonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR5375-100G OR5375-25G OR5375-5G
76621-586EA 124.28 USD
76621-586 76621-588 76621-590
3-Chlorobenzenesulfonyl chloride 98%
3-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 102…104 °C/1mmHg
Melting Pt: 21 °C
Density: 1.499 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051697
CAS Number: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR