Sulfur trioxide pyridine complex
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR24145-100G
OR24145-500G
76620-842EA
38.72
USD
76620-842
76620-844
Sulfur trioxide pyridine complex
Sulphur trioxide pyridine complex
Formula:
C₅H₅NO₃S Melting Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012437 CAS Number: 26412-87-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...