VWR® DNA Ladders

Supplier: VWR International

76620-562 76620-564 76620-566
76620-562EA 49.76 USD
76620-562 76620-564 76620-566
VWR® DNA Ladders
Electrophoresis Reagents Molecular Weight Markers

DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.


  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose


Mini DNA Ladder molecular range: 100 to 500 bp

Low DNA Ladder molecular range: 100 to 1000 bp

PCR Ladder molecular range: 100 to 3000 bp

Mini DNA Ladder, Low DNA Ladder and PCR Ladder are supplied in 0.5 mL packs for 100 lanes.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR