VWR® DNA Ladders

Supplier: VWR International

76620-556 76620-558 76620-560
76620-556EA 305.89 USD
76620-556 76620-558 76620-560
VWR® DNA Ladders
Electrophoresis Reagents Molecular Weight Markers

DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.


  • Supplied in ready to use loading buffer
  • For direct loading and easy visualisation
  • Clear, distinct bands
  • Mass-calibrated bands for DNA quantification
  • PCR DNA LADDER has a 1000 bp band that is extra bright to serve as reference point
  • Blue dye front running at 100 to 300 bp at 0.5 to 1.5% agarose


High Range DNA Ladder molecular range: 200 to 12000 bp; mass-calibrated bands from 6 to 40 ng/band for DNA quantification.

Low Range DNA Ladder molecular range: 100 to 1000 bp; mass-calibrated bands from 8 to 40 ng/band for DNA quantification.

PCR DNA Ladder molecular range: 100 to 3000 bp; mass-calibrated bands from 10 to 30 ng/band for DNA quantification.

High Range DNA Ladder, Low Range DNA Ladder and PCR DNA Ladder are supplied in 0.5 mL packs sufficient for 250 lanes.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR