Water, PCR grade

Supplier: VWR International

76620-532
76620-532EA 44.37 USD
76620-532
Water, PCR grade
Water

Ultrapure water for PCR


  • Convenient 5 mL aliquots
  • Tested for contaminating activities


PCR Grade Water is ultrapure H2O, tested for contaminating activities, with no trace of endonuclease activity, nicking activity or exonuclease activity. Absence of human DNA is tested by PCR.

Storage at room temperature. Long term storage at -20 °C.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

DNA nicking activity Passed
Endonuclease activity Passed
Exonuclease activity Passed
Absence of Human DNA Passed
Volume check, > 5 ml/tube Passed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR