VWR® Glycerol-Free Taq DNA Polymerase
Supplier: VWR International
76620-416 76620-418 76620-420 76620-422
76620-416EA 689.93 USD
76620-416 76620-418 76620-420 76620-422
VWR® Glycerol-Free Taq DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq DNA Polymerase, glycerol-free, 5 U/µl is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.
  • Glycerol-free storage buffer
  • Suitable for automated high-throughput testing
  • Ideal for freeze drying protocols
  • Suitable for robot-aided pipetting
  • Processes up to 5 kb
  • dUTP incorporation possible
  • Leaves a 3'dA overhang
Taq DNA Polymerase, glycerol-free, concentration: 5 Units/μl
Taq DNA Polymerase, glycerol-free, has both a 5'→3' DNA polymerase and a 5'→3' exonuclease activity. The enzyme lacks a 3'→5' exonuclease activity (no proofreading ability). VWR® Taq DNA Polymerase, glycerol-free, leaves an A' overhang, which makes the enzyme ideal for TA cloning.
Long term storage at -20 °C. Product expiry at -20 °C is stated on the label. Optional: Store at +4 °C for up to 6 months.
Caution: For Research Use Only. Not for use in diagnostic procedures.