(2S)-2-Decanamido-3-methylbutanoic acid 95%

Supplier: AFG Bioscience

C73078-100MG C73078-200MG
76620-404EA 450.69 USD
76620-404 76620-406
(2S)-2-Decanamido-3-methylbutanoic acid 95%
(2S)-2-Decanamido-3-methylbutanoic acid
Formula: C₁₅H₂₉NO₃
Storage Temperature: Refrigerator
CAS Number: 22220-07-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR