DNP-PEG3-amine ≥95%
Supplier: AFG Bioscience
C59832-1G
C59832-2G
76620-382EA
550.84
USD
76620-382
76620-384
DNP-PEG3-amine ≥95%
DNP-PEG3-amine
Formula:
C₁₂H₁₈N₄O₆ Storage Temperature: Refrigerator |
CAS Number:
929521-54-0 |
Specification Test Results
Purity Limit | > 95% |
Appearance | sticky oil |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...