Fmoc-Val-Cit-PAB-PNP 95%

Supplier: AFG Bioscience

C56028-100MG C56028-250MG C56028-500MG
76620-216EA 98.58 USD
76620-216 76620-218 76620-220
Fmoc-Val-Cit-PAB-PNP 95%
Fmoc-Val-Cit-PAB-PNP
Formula: C₄₀H₄₂N₆O₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD22200278
CAS Number: 863971-53-3

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR