3-O-Methyl-N-acetyl-D-glucosamine ≥95%

Supplier: AFG Bioscience

Synonyms: 3-OMe-GlcNAc, 2-​Acetamido-​2-​deoxy-​3-​O-​methyl-​D-​glucopyranose

C54109-10MG C54109-25MG C54109-50MG
76620-028EA 150.23 USD
76620-028 76620-030 76620-032
3-O-Methyl-N-acetyl-D-glucosamine ≥95%
3-O-Methyl-N-acetyl-D-glucosamine
Formula: C₉H₁₇NO₆
Storage Temperature: Freezer
CAS Number: 94825-74-8

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR