N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid ≥95%

Supplier: AFG Bioscience

Synonyms: N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid, 2,3-Dehydro-2-deoxy-N-acetylneuraminic acid, DANA

C53830-100MG C53830-25MG C53830-10MG
76619-974EA 901.37 USD
76619-974 76619-972 76619-970
N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid ≥95%
N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid
Formula: C₁₁H₁₇NO₈
Storage Temperature: Freezer
MDL Number: MFCD00057470
CAS Number: 24967-27-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR