2-O-Methyl-α-D-N-acetylneuraminic acid ≥95%
Supplier: AFG Bioscience
Synonyms:
N-Acetyl-2-O-methyl-a-D-neuraminic acid
C53621-100MG
C53621-25MG
C53621-50MG
C53621-10MG
76619-940EA
845.81
USD
76619-940
76619-936
76619-938
77888-927
2-O-Methyl-α-D-N-acetylneuraminic acid ≥95%
2-O-Methyl-α-D-N-acetylneuraminic acid
Formula:
C₁₂H₂₁NO₉ Storage Temperature: Freezer |
MDL Number:
MFCD00084809 CAS Number: 50930-22-8 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White To Off-White Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...