L(-)-Galactose 98%

Supplier: AFG Bioscience

Synonyms: (2S,3R,4R,5S)-2,3,4,5,6-Pentahydroxyhexanal, (2S,3R,4R,5S)-Pentahydroxyhexanal, L-Galactose

C53477-1G C53477-250MG C53477-500MG
76619-916EA 450.69 USD
76619-916 76619-912 76619-914
L(-)-Galactose 98%
L(-)-Galactose
Formula: C₆H₁₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00063833
CAS Number: 15572-79-9

Order Now

Specification Test Results

Purity Limit 98%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR