Phenylethyl-β-D-thiogalactopyranoside ≥98%
Supplier: AFG Bioscience
Synonyms:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenethylthio)tetrahydro-2H-pyran-3,4,5-triol
C53297-1G
C53297-2G
C53297-500MG
76619-886EA
299.67
USD
76619-886
76619-888
76619-884
Phenylethyl-β-D-thiogalactopyranoside ≥98%
Phenylethyl-β-D-thiogalactopyranoside
Formula:
C₁₄H₂₀O₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00467904 CAS Number: 63407-54-5 |
Specification Test Results
Purity Limit | > 98% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...