β-Glc-PEG3-Amine 95%

Supplier: AFG Bioscience

C53009-25MG C53009-50MG
76619-840EA 250.38 USD
76619-840 76619-842
β-Glc-PEG3-Amine 95%
β-Glc-PEG3-Amine
Formula: C₁₂H₂₅NO₈
Storage Temperature: Freezer
CAS Number: 928850-73-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless sticky syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR