Propargyl α-D-mannopyranoside 95%

Supplier: AFG Bioscience

Synonyms: 2-Propyn-1-yl α-D-mannoside, Propargyl ALPHA-D-mannopyranoside, 2-Propynyl α-D-mannopyranoside

C52012-100MG C52012-250MG
76619-752EA 200.3 USD
76619-752 76619-754
Propargyl α-D-mannopyranoside 95%
Propargyl α-D-mannopyranoside
Formula: C₉H₁₄O₆
Storage Temperature: Freezer
MDL Number: MFCD19980990
CAS Number: 854262-01-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR