β-GlcNAc-PEG3-Alkyne 95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-β-D-glucopyranoside, N-((2R,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(2-(2-(2-(prop-2-yn-1-yloxy)ethoxy)ethoxy)ethoxy)tetrahydro-2H-pyran-3-yl)acetamide
C52002-25MG
C52002-50MG
76619-720EA
295.75
USD
76619-720
76619-722
β-GlcNAc-PEG3-Alkyne 95%
β-GlcNAc-PEG3-Alkyne
Formula:
C₁₇H₂₉NO₉ MW: 391.41 g/mol Storage Temperature: Freezer |
CAS Number:
1884184-44-4 |
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...