α-GalNAc-PEG3-alkyne 95%
Supplier: AFG Bioscience
Synonyms:
2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-α-D-galactopyranoside
C52001-25MG
C52001-50MG
C52001-100MG
C52001-10MG
76619-716EA
450.69
USD
76619-716
76619-718
77888-823
77888-824
α-GalNAc-PEG3-alkyne 95%
α-GalNAc-PEG3-alkyne
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...