2-Azidoethyl LacNAc ≥95%

Supplier: AFG Bioscience

Synonyms: 2-Azidoethyl N-acetyl-D-lactosamine

C51783-25MG C51783-10MG C51783-2MG C51783-5MG
76619-694EA 1490.53 USD
76619-694 76619-692 77888-816 77888-817
2-Azidoethyl LacNAc ≥95%
2-Azidoethyl LacNAc
Formula: C₁₆H₂₈N₄O₁₁
Storage Temperature: Freezer
MDL Number: MFCD19981037
CAS Number: 338971-38-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR