2,​3,​6,​2',​3',​4',​6'-​Hepta-​O-​acetyl-​b-​maltosyl azide 95%

Supplier: AFG Bioscience

Synonyms: 2,3,6-Tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-a-D-glucopyranosyl)-b-D-glucopyranosyl azide, b-Maltosylazide heptaacetate

C51762-10G C51762-1G C51762-5G C51762-100MG C51762-250MG C51762-2G
76619-672EA 1498.37 USD
76619-672 76619-668 76619-670 77888-808 77888-809 77888-810
2,​3,​6,​2',​3',​4',​6'-​Hepta-​O-​acetyl-​b-​maltosyl azide 95%
2,​3,​6,​2',​3',​4',​6'-​Hepta-​O-​acetyl-​b-​maltosyl azide
Formula: C₂₆H₃₅N₃O₁₇
Storage Temperature: Refrigerator
MDL Number: MFCD00216969
CAS Number: 33012-49-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance Off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR