3-​Azidopropyl 2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranoside ≥95%

Supplier: AFG Bioscience

C51578-1G C51578-250MG C51578-500MG C51578-100MG C51578-25MG
76619-616EA 1348.92 USD
76619-616 76619-612 76619-614 77888-758 77888-759
3-​Azidopropyl 2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranoside ≥95%
3-​Azidopropyl 2-​Acetamido-​2-​deoxy-​β-​D-​glucopyranoside
Formula: C₁₁H₂₀N₄O₆
Storage Temperature: Refrigerator
CAS Number: 595568-99-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR