2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside 95%

Supplier: AFG Bioscience

Synonyms: β-D-GalNAc-Et-N3

C51573-100MG C51573-250MG C51573-25MG
76619-608EA 500.76 USD
76619-608 76619-610 76619-606
2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside 95%
2-Azidoethyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside
Formula: C₁₀H₁₈N₄O₆
Storage Temperature: Freezer
MDL Number: MFCD19981043
CAS Number: 142072-15-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR