2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide 97%

Supplier: AFG Bioscience

Synonyms: 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl Azide, 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate

C51570-1G C51570-2G C51570-5G
76619-594EA 120.18 USD
76619-594 76619-596 76619-598
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide 97%
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
Formula: C₁₄H₂₀N₄O₈
Storage Temperature: Refrigerator
MDL Number: MFCD00216968
CAS Number: 6205-69-2

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR