2-acetylamino-6-azido-2,6-dideoxy-D-galactose ≥95%
Supplier: AFG BIOSCIENCE LLC
C51525-100MG
C51525-50MG
76619-492EA
640.8
USD
76619-492
76619-490
2-acetylamino-6-azido-2,6-dideoxy-D-galactose ≥95%
2-acetylamino-6-azido-2,6-dideoxy-D-galactose
Formula:
C₈H₁₄N₄O₅ MW: 246.22 g/mol Storage Temperature: Freezer |
CAS Number:
1190619-44-3 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...