1-Azido-1-deoxy-β-D-galactopyranoside 97%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
β-D-Galactopyranosyl azide
C51522-1G
C51522-2G
C51522-500MG
76619-480EA
499.58
USD
76619-480
76619-482
76619-478
1-Azido-1-deoxy-β-D-galactopyranoside 97%
1-Azido-1-deoxy-β-D-galactopyranoside
Formula:
C₆H₁₁N₃O₅ MW: 205.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00181720 CAS Number: 35899-89-9 |
Specification Test Results
Purity Limit | 97% |
Appearance | White Crystalline Solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...