1-Azido-1-deoxy-β-D-galactopyranoside 97%

Supplier: AFG BIOSCIENCE LLC

Synonyms: β-D-Galactopyranosyl azide

C51522-1G C51522-2G C51522-500MG
76619-480EA 499.58 USD
76619-480 76619-482 76619-478
1-Azido-1-deoxy-β-D-galactopyranoside 97%
1-Azido-1-deoxy-β-D-galactopyranoside
Formula: C₆H₁₁N₃O₅
MW: 205.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00181720
CAS Number: 35899-89-9

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR