2-Azidoethyl-β-D-glucopyranoside 95%

Supplier: AFG Bioscience

Synonyms: β-D-Glc-Et-N3

C51511-1G C51511-500MG C51511-250MG
76619-434EA 591.51 USD
76619-434 76619-432 76619-430
2-Azidoethyl-β-D-glucopyranoside 95%
2-Azidoethyl-beta-D-glucopyranoside
Formula: C₈H₁₅N₃O₆
Storage Temperature: Freezer
CAS Number: 165331-08-8

Order Now

Specification Test Results

Purity Limit 95%
Appearance Colorless Syrup


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR