2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine ≥95%

Supplier: AFG Bioscience

Synonyms: β-D-GlcNAc-(1→N)-Asn, Ng-(2-Acetamido-2-deoxy-b-D-gluco-pyranosyl)-L-asparagine, 2-Acetamido-1-β-(L-aspartamido)-1 ,2-dideoxy-D-glucose, H-Asn(GlcNAc-β-D)-OH

C51064-100MG C51064-250MG C51064-25MG
76619-364EA 849.73 USD
76619-364 76619-366 76619-362
2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine ≥95%
2-​Acetamido-​2-​deoxy-​​β-​D-​glucopyranosyl L-​asparagine
Formula: C₁₂H₂₁N₃O₈
Storage Temperature: Refrigerator
MDL Number: MFCD02259695
CAS Number: 2776-93-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance Off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR