Fmoc-L-Thr (β-D-Glc(Ac)4)-OH 95%

Supplier: AFG Bioscience

Synonyms: N-α-(9-Fluorenylmethyloxycarbonyl)-O-β-(2,3,4,6-tetra-O-acetyl-β-D-glucopyranosyl)-L-threonine

C51029-100MG C51029-250MG C51029-25MG
76619-340EA 394.34 USD
76619-340 76619-342 76619-338
Fmoc-L-Thr (β-D-Glc(Ac)4)-OH 95%
Fmoc-L-Thr (β-D-Glc(Ac)4)-OH
Formula: C₃₃H₃₇NO₁₄
Storage Temperature: Freezer
CAS Number: 130548-92-4

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR