Fmoc-L-Asn(GlcNAc(Ac)3-β)-OH ≥95%

Supplier: AFG Bioscience

Synonyms: Fmoc-Asn(GlcNAc(Ac)₃-β-D)-OH, N-α-(Fluorenylmethoxycarbonyl)-N-γ-(2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl)-L-asparagine, Fmoc-L-Asn((Ac)3-β-D-GlcNAc)-OH

C51001-100MG C51001-250MG C51001-500MG
76619-254EA 250.38 USD
76619-254 76619-256 76619-258
Fmoc-L-Asn(GlcNAc(Ac)3-β)-OH ≥95%
Fmoc-L-Asn(GlcNAc(Ac)3-beta)-OH
Formula: C₃₃H₃₇N₃O₁₃
Storage Temperature: Freezer
MDL Number: MFCD00237079
CAS Number: 131287-39-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR