Mannose triflate ≥95%

Supplier: AFG Bioscience

Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, TATM

C50648-500MG C50648-1G C50648-2G
76619-110EA 150.23 USD
76619-110 76619-112 76619-114
Mannose triflate ≥95%
Mannose triflate
Formula: C₁₅H₁₉F₃O₁₂S
Storage Temperature: Freezer
MDL Number: MFCD00012353
CAS Number: 92051-23-5

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR