Mannose triflate ≥95%
Supplier: AFG Bioscience
Synonyms:
(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, TATM
C50648-500MG
C50648-1G
C50648-2G
76619-110EA
150.23
USD
76619-110
76619-112
76619-114
Mannose triflate ≥95%
Mannose triflate
Formula:
C₁₅H₁₉F₃O₁₂S Storage Temperature: Freezer |
MDL Number:
MFCD00012353 CAS Number: 92051-23-5 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...