3',4',5'-Trifluoropropiophenone 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7820EA-25G
PC7820EA-5G
PC7820EA-1G
76618-928EA
347.4
USD
76618-928
76618-930
76618-926
3',4',5'-Trifluoropropiophenone 97%
3',4',5'-Trifluoropropiophenone
Formula:
C₉H₇F₃O MW: 188.15 Storage Temperature: Ambient |
CAS Number:
220227-74-7 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...