7-Hydroxy-4-(trifluoromethyl)coumarin 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-(Trifluoromethyl)umbelliferone, TFMU
PC4866-1G
PC4866-5G
76618-396EA
22.42
USD
76618-396
76618-398
7-Hydroxy-4-(trifluoromethyl)coumarin 97%
7-Hydroxy-4-(trifluoromethyl)coumarin
Formula:
C₁₀H₅F₃O₃ Melting Pt: 178…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037578 CAS Number: 575-03-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...