4-Nitro-2-(trifluoromethyl)aniline 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC1080-100G PC1080-25G
76617-684EA 41.76 USD
76617-684 76617-686 76617-688
4-Nitro-2-(trifluoromethyl)aniline 95%
4-Nitro-2-(trifluoromethyl)aniline
Formula: C₇H₅F₃N₂O₂
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR