1-Boc-4-(aminomethyl)piperidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

OR8043-1G OR8043-5G
76617-348EA 20.87 USD
76617-348 76617-350 76617-352
1-Boc-4-(aminomethyl)piperidine 97%
1-Boc-4-(aminomethyl)piperidine
Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 237…238 °C/760mmHg
Density: 1.013 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01076207
CAS Number: 144222-22-0
UN: 3082
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR