1-Benzyl-4-hydroxypiperidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Benzylpiperidin-4-ol

OR6935-500G
76617-258EA 129.78 USD
76617-258 76617-260
1-Benzyl-4-hydroxypiperidine 98%
1-Benzyl-4-hydroxypiperidine
Formula: C₁₂H₁₇NO
Boiling Pt: 127…128 °C/2mmHg
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006503
CAS Number: 4727-72-4
EINECS: 225-226-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR