1-(2-Methoxyphenyl)-2-thiourea
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea
OR5634-1G
OR5634-5G
76617-032EA
20.87
USD
76617-032
76617-034
1-(2-Methoxyphenyl)-2-thiourea
1-(2-Methoxyphenyl)-2-thiourea
Formula:
(C₆H₄(OCH₃))NC(S)NH₂ Storage Temperature: Ambient |
MDL Number:
MFCD00022162 CAS Number: 1516-37-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...