6-Thioguanine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR4016-1G
OR4016-5G
76616-678EA
20.28
USD
76616-678
76616-680
6-Thioguanine
6-Thioguanine
Formula:
C₅H₅N₅S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233553 CAS Number: 154-42-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...