6-Chloro-7-deazapurine

Supplier: APOLLO SCIENTIFIC
Danger

OR2836-25G OR2836-5G
76616-222EA 28.52 USD
76616-222 76616-224
6-Chloro-7-deazapurine
6-Chloro-7-deazapurine
Formula: C₆H₄ClN₃
Melting Pt: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD01686865
CAS Number: 3680-69-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR