Indole
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1H-Indole
OR10189-100G
OR10189-25G
OR10189-500G
76615-308EA
41.76
USD
76615-308
76615-310
76615-312
Indole
Indole
Formula:
C₈H₇N MW: 117.15 g/mol Boiling Pt: 253…254 °C Melting Pt: 52…54 °C Density: 1.22 g/cm³ (25 °C) Flash Pt: 121 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005607 CAS Number: 120-72-9 EINECS: 204-420-7 UN: 2811 ADR: 6.1 III Merck Index: 13,04985 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...